Would you like to log in again?
John Guest acetal push-to-connect union T-connectors
Supplier: John Guest
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PM0208S PI0208S-US PI0206S PP0208W-US PI0212S-US PI0216S PP0212W-US PM0204S
MFLX34006-53EA 47.2 USD InStock
MFLX34006-53 MFLX34006-30 MFLX34006-11 MFLX34007-46 MFLX34006-54 MFLX34007-47 MFLX34006-13 MFLX34006-12
John Guest acetal push-to-connect union T-connectors
Fittings and Connectors
No need to buy two fitting halves.
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth— tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty