Would you like to log in again?
John Guest push-to-connect to male NPT adapters
Supplier: John Guest
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PP010821W PI011222S-US PP010822W-US PP011223W PI011224S PI011223S-US PI011624S PI011221S PM010822S PM010421S PI010822S PM010821S PI011623S PI010823S PI010821S PI010621S PM010823S PP011222W PM010422S
MFLX34007-42EA 22.3 USD InStock
MFLX34007-42 MFLX34006-51 MFLX34007-43 MFLX34006-50 MFLX34006-52 MFLX34006-02 MFLX34007-44 MFLX34007-45 MFLX34006-04 MFLX34006-48 MFLX34006-03 MFLX34006-06 MFLX34006-28 MFLX34006-05 MFLX34006-49 MFLX34006-08 MFLX34006-07 MFLX34006-29 MFLX34006-09
John Guest push-to-connect to male NPT adapters
Fittings and Connectors
No need to buy two fitting halves.
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth— tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty