Would you like to log in again?
John Guest Acetal Push-To-Connect Male Threaded Adapter Fittings
Supplier: John Guest
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PM050822S PM050823S PI051223S PM050421S PM050821S PI050822S PI050621S PM050422S PI051623S PI051222S PI051624S PI050821S
MFLX34007-28EA 13 USD InStock
MFLX34007-28 MFLX34006-69 MFLX34007-29 MFLX34007-27 MFLX34006-71 MFLX34006-70 MFLX34007-31 MFLX34006-86 MFLX34007-30 MFLX34006-66 MFLX34006-68 MFLX34006-67
John Guest Acetal Push-To-Connect Male Threaded Adapter Fittings
Fittings and Connectors
No need to buy two fitting halves.
  • Meets NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
  • A single fitting offers secure connections
  • and quick disconnection
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty