Would you like to log in again?
John Guest Polypropylene Push-To-Connect Elbow Adapter Fittings
Supplier: John Guest
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PP221208W PP221212W-US PP220808W-US
MFLX34007-59EA 48 USD InStock
MFLX34007-59 MFLX34007-60 MFLX34007-58
John Guest Polypropylene Push-To-Connect Elbow Adapter Fittings
Fittings and Connectors
Quick, leakproof connection.
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters let you permanently modify basic fittings for customized installations.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty