Would you like to log in again?
Union Fittings, Female NPT, 90° Elbow, PTFE
Supplier: MICROMOLD PRODUCTS, INC.
F-90E08 F-90E04 F-90E01 F-90E02
MFLX34030-07EA 56.8 USD InStock
MFLX34030-07 MFLX34030-08 MFLX34030-10 MFLX34030-12
Union Fittings, Female NPT, 90° Elbow, PTFE
Fittings and Connectors
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty