Would you like to log in again?
Masterflex® 1½ Sanitary Tri-Clamp Cap", Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SF16-CAP-PP-QC-MF 10PK SF16-CAP-CVU-QC-MF 10PK SF16-CAP-NK7-QC-MF 10PK
MFLX40330-21EA 293 USD InStock
MFLX40330-21 MFLX40330-20 MFLX40330-19
Masterflex® 1½ Sanitary Tri-Clamp Cap", Avantor®
Fittings and Connectors
Close off your sanitary fluid line with a secure connection.
  • Fully compatible with other similar sized Tri-Clamp® connections
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Sterilizable via gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. These adapters offer good chemical resistance and may be sterilized by gamma irradiation. Clamps and gaskets required for a complete system are sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty