Would you like to log in again?
Masterflex® Cleanroom 1 Sanitary Flange to Hose Barb", Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SF16-SB12-CVU-QC-MF 10PK SF16-SB16-NK7-QC-MF 10PK SF16-SB4-NK7-QC-MF 10PK SF16-SB4-PP-QC-MF 10PK SF16-SB8-NK7-QC-MF 10PK SF16-SB12-NK7-QC-MF 10PK SF16-SB12-PS-QC-MF 10PK SF16-SB6-NK7-QC-MF 10PK SF16-SB6-PP-QC-MF 10PK SF16-SB16-PP-QC-MF 10PK SF16-SB16-CVU-QC-MF 10PK SF16-SB8-PP-QC-MF 10PK SF16-SB8-CVU-QC-MF 10PK SF16-SB12-PP-QC-MF 10PK SF16-SB6-CVU-QC-MF 10PK SF16-SB8-PS-QC-MF 10PK SF16-SB4-PS-QC-MF 10PK SF16-SB6-PS-QC-MF 10PK SF16-SB16-PS-QC-MF 10PK SF16-SB4-CVU-QC-MF 10PK
MFLX40330-23EA 196 USD InStock
MFLX40330-23 MFLX40330-34 MFLX40330-22 MFLX40330-33 MFLX40330-32 MFLX40330-31 MFLX40330-41 MFLX40330-30 MFLX40330-40 MFLX40330-29 MFLX40330-39 MFLX40330-28 MFLX40330-27 MFLX40330-38 MFLX40330-26 MFLX40330-37 MFLX40330-25 MFLX40330-36 MFLX40330-35 MFLX40330-24
Masterflex® Cleanroom 1 Sanitary Flange to Hose Barb", Avantor®
Fittings and Connectors
Maintain sanitary conditions with secure connections from sanitary Tri-Clamp to tubing.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
  • Compatible with other Tri-Clamp® connections
The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use industries. They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave. The PVDF fittings withstand higher temperatures than the Polypropylene fittings. Clamps and gaskets required for a complete system are sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty