Would you like to log in again?
Masterflex® Single-Use Bag Port Disc Opposing Barb
Supplier: Avantor Fluid Handling
Masterflex®
PDO4-E8402-QC-MF-10PK PDO3-E8402-QC-MF-10PK PDO8-E8402-QC-MF-10PK PDO2-E8402-QC-MF-10PK PDO6-E8402-QC-MF-10PK
MFLX40331-14EA 38 USD InStock
MFLX40331-14 MFLX40331-15 MFLX40331-11 MFLX40331-12 MFLX40331-13
Masterflex® Single-Use Bag Port Disc Opposing Barb
Fittings and Connectors
Designed for sample port and dip tube applications.
  • Barb sizes range from 1/8" to 1/2"
  • 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty