Would you like to log in again?
Masterflex® Single-Use Bag Port Disc Flat Plate
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
PND4-E8402-QC-MF-10PK PND3-E8402-QC-MF-10PK PND45-6-E8402-MF-10PK PND10-E8402-QC-MF-10PK PND2.5-E8402-QC-MF-10PK PND8-E8402-QC-MF-10PK PND6-E8402-QC-MF-10PK PND12-E8402-QC-MF-10PK PND-LF E8402-QC-MF-10PK PND2-E8402-QC-MF-10PK PND5-E8402-QC-MF-10PK PND16-E8402-QC-MF-10PK
MFLX40331-25EA 35.5 USD InStock
MFLX40331-25 MFLX40331-26 MFLX40331-16 MFLX40331-27 MFLX40331-17 MFLX40331-18 MFLX40331-19 MFLX40331-20 MFLX40331-21 MFLX40331-22 MFLX40331-23 MFLX40331-24
Masterflex® Single-Use Bag Port Disc Flat Plate
Fittings and Connectors
Flexible, smooth flat base.
  • Barb sizes range from 1/8" to 1"
  • 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty