Would you like to log in again?
Masterflex® Single-Use Bag Port Plate Ribbed Base, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
PP2.5-E8402-QC-MF-10PK PP3-E8402-QC-MF-10PK PP2-E8402-QC-MF-10PK PP12-E8402-QC-MF-10PK PP10-E8402-QC-MF-10PK PP4-E8402-QC-MF-10PK PP16-E8402-QC-MF-10PK PP6-E8402-QC-MF-10PK PP5-E8402-QC-MF-10PK PP8-E8402-QC-MF-10PK
MFLX40332-04EA 33.1 USD InStock
MFLX40332-04 MFLX40332-05 MFLX40332-06 MFLX40332-07 MFLX40332-08 MFLX40332-09 MFLX40332-00 MFLX40332-01 MFLX40332-02 MFLX40332-03
Masterflex® Single-Use Bag Port Plate Ribbed Base, Avantor®
Fittings and Connectors
Ribbed base prevents suction blockage and increases rigidity.
  • Barb sizes range from 1/8" to 1"
  • 1-3/4" and 2-1/4" ribbed bases
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty