Would you like to log in again?
Masterflex® Adapter Fittings, Hose Barb to Threaded Adapter, Straight, PVDF, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
A4-8NK7-MF 10PK A12-10NK7-MF 10PK A6-5NK7-MF 10PK A4-2.5NK7-MF 10PK A2-2NK7-MF 10PK A4-5NK7-MF 10PK A8-10NK7-MF 10PK A8-10NK7-MF 10PK A2-2NK7 A2-6NK7-MF 10PK A6-4NK7-MF 10PK A12-12NK7-MF 10PK A2-5NK7-MF 10PK A6-6NK7-MF 10PK A8-6NK7-MF 10PK A2-4NK7-MF 10PK A4-6NK7-MF 10PK A2-3NK7-MF 10PK A8-8NK7-MF 10PK A6-8NK7-MF 10PK A8-12NK7-MF 10PK A12-4NK7-MF 10PK A16-16NK7-MF 10PK A12-16NK7-MF 10PK A4-3NK7-MF 10PK A4-2NK7-MF 10PK A4-4NK7-MF 10PK A8-12NK7-MF 10PK A8-4NK7-MF 10PK A12-6NK7-MF 10PK
MFLX40614-32EA 79.2 USD InStock
MFLX40614-32 MFLX40614-10 MFLX40614-33 MFLX40614-11 MFLX40614-12 MFLX40614-34 MFLX40614-35 MFLX40614-13 MFLX40614-30 MFLX40614-31 MFLX40614-18 MFLX40614-19 MFLX40614-14 MFLX40614-15 MFLX40614-16 MFLX40614-17 MFLX40614-21 MFLX40614-22 MFLX40614-23 MFLX30704-07 MFLX40614-24 MFLX40614-20 MFLX40614-07 MFLX40614-29 MFLX40614-08 MFLX40614-09 MFLX40614-25 MFLX40614-26 MFLX40614-27 MFLX40614-28
Masterflex® Adapter Fittings, Hose Barb to Threaded Adapter, Straight, PVDF, Avantor®
Fittings and Connectors
Quick and easy installation with no tools required.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). PVDF material features excellent chemical resistance with a wide temperature range. Sterilize via ethylene oxide or autoclave.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty