Would you like to log in again?
Masterflex® Reducer Fittings, Hose Barb, Straight (PVDF), Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
C10-8NK7-MF 10PK C6-4NK7-MF 10PK C3-1-200NK7-MF 10PK C6-4-AGNK7-MF 10PK C12-10NK7-MF 10PK C4-2NK7-MF 10PK C2-1-200NK7-MF 10PK C8-6NK(10 PACK) C2.5-2NK7-MF 10PK C10-6NK7-MF 10PK C8-6NK7-MF 10PK C4-3NK7-MF 10PK C10-4NK7-MF 10PK C5-4NK7-MF 10PK C16-12NK7-MF 10PK C8-5NK7-MF 10PK C10-5NK7-MF 10PK C6-5NK7-MF 10PK C12-8NK7-MF 10PK C1.5-1-200NK7-MF 10PK C4-2.5NK7-MF 10PK C6-2NK7-MF 10PK C8-4NK7-MF 10PK C2-1.5-200NK7-MF 10PK C16-8NK7-MF 10PK C16-10NK7-MF 10PK C12-6NK7-MF 10PK C3-2NK7-MF 10PK
MFLX40614-70EA 53.5 USD InStock
MFLX40614-70 MFLX40614-71 MFLX40614-76 MFLX40614-77 MFLX40614-55 MFLX40614-56 MFLX40614-78 MFLX40614-57 MFLX40614-72 MFLX40614-51 MFLX40614-73 MFLX40614-52 MFLX40614-74 MFLX40614-53 MFLX40614-75 MFLX40614-58 MFLX40614-59 MFLX40614-60 MFLX40614-65 MFLX30703-58 MFLX40614-66 MFLX40614-67 MFLX40614-68 MFLX40614-61 MFLX40614-62 MFLX40614-63 MFLX40614-64 MFLX40614-69
Masterflex® Reducer Fittings, Hose Barb, Straight (PVDF), Avantor®
Fittings and Connectors
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range -80 to 265 °F (-62 to 129 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty