Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Tee, PVDF, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
L6-4NK7-MF 10PK L2-6NK7-MF 10PK L4-2NK7-MF 10PK L2-5NK7-MF 10PK L12-6NK7-MF 10PK L4-6NK7-MF 10PK L4-8NK7-MF 10PK L8-12NK7-MF 10PK L2-2NK7-MF 10PK L2-4NK7-MF 10PK L12-12NK7-MF 10PK L12-4NK7-MF 10PK L12-8NK7-MF 10PK L4-3NK7-MF 10PK L4-4NK7-MF 10PK L8-8NK7-MF 10PK L6-6NK7-MF 10PK L6-8NK7-MF 10PK L8-4NK7-MF 10PK L8-6NK7-MF 10PK
MFLX40615-00EA 73.2 USD InStock
MFLX40615-00 MFLX40615-11 MFLX40615-01 MFLX40615-12 MFLX40615-13 MFLX40615-02 MFLX40615-14 MFLX40615-03 MFLX40615-20 MFLX40615-08 MFLX40615-19 MFLX40615-09 MFLX40615-04 MFLX40615-15 MFLX40615-05 MFLX40615-16 MFLX40615-06 MFLX40615-17 MFLX40615-07 MFLX40615-18
Masterflex® Union Fittings, Hose Barb, Tee, PVDF, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty