Would you like to log in again?
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, PVDF, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
T8-6NK7-MF 10PK T6-8NK7-MF 10PK T4-6NK7-MF 10PK T2-6NK7-MF 10PK T6-4NK7-MF 10PK T8-8NK7-MF 10PK T8-4NK7-MF 10PK T6-6NK7-MF 10PK T4-4NK7-MF 10PK T8-12NK7-MF 10PK T12-4NK7-MF 10PK T2-4NK7-MF 10PK T4-8NK7-MF 10PK
MFLX40615-80EA 79.2 USD InStock
MFLX40615-80 MFLX40615-70 MFLX40615-71 MFLX40615-72 MFLX40615-77 MFLX40615-78 MFLX40615-79 MFLX40615-68 MFLX40615-69 MFLX40615-73 MFLX40615-74 MFLX40615-75 MFLX40615-76
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, PVDF, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty