Would you like to log in again?
Masterflex® Union Fittings, Sanitary Clamp, Tee, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
TT250 KT100L TT150 TT075 KT075 KT100L KT150 KT150 TT200 TT100L TT300 KT200 KT200 KT075
MFLX31806-61EA 91.1 USD InStock
MFLX31806-61 MFLX40618-73 MFLX40618-72 MFLX31806-63 MFLX31809-64 MFLX31809-63 MFLX31806-62 MFLX31809-62 MFLX31809-61 MFLX40618-71 MFLX40618-70 MFLX31806-65 MFLX31806-64 MFLX31806-66
Masterflex® Union Fittings, Sanitary Clamp, Tee, Avantor®
Fittings and Connectors
Maintain sanitary conditions while combining or dividing flow.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
Tri-Clamp® standard tee adapters allow for liquid flow to be dividing into two or combined into one flow path.
Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving.
Clamps and gaskets required for a complete system are sold separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty