Would you like to log in again?
Masterflex® Adapter Fittings, Sanitary Clamp to Male Thread, Straight, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
TAD100L-06M KAD100L-04M KAD200-20M KAD075-08M KAD100L-16M KAD150-24M TAD100L-04M KAD150-12M TAD200-32M KAD100L-08M TAD150-24M TAD075-08M TAD150-20M KAD075-06M KAD100L-12M TAD100L-08M KAD100L-20M KAD200-32M TAD150-12M KAD075-04M KAD100L-16M KAD150-24M KAD200-16M TAD075-04M KAD150-20M KAD150-12M KAD075-04M TAD100L-16M TAD100L-12M TAD200-16M TAD250-32M KAD150-16M KAD200-24M TAD100L-24M KAD100L-06M TAD200-24M KAD100L-08M TAD150-16M KAD075-08M TAD075-02M TAD075-06M
MFLX31200-03EA 148 USD InStock
MFLX31200-03 MFLX31200-02 MFLX40619-42 MFLX31805-60 MFLX40619-40 MFLX40619-41 MFLX31805-61 MFLX31808-71 MFLX31805-80 MFLX31805-68 MFLX31805-89 MFLX31808-59 MFLX40619-49 MFLX31805-69 MFLX40619-46 MFLX40619-47 MFLX31805-63 MFLX31808-77 MFLX40619-44 MFLX40619-45 MFLX31201-01 MFLX31201-03 MFLX31805-71 MFLX31805-70 MFLX40619-51 MFLX31808-61 MFLX40619-50 MFLX31805-57 MFLX40619-39 MFLX31805-79 MFLX40619-37 MFLX31805-59 MFLX31805-58 MFLX40619-38 MFLX31808-68 MFLX40619-35 MFLX40619-36 MFLX31805-77 MFLX40619-34 MFLX31200-01 MFLX31200-00
Masterflex® Adapter Fittings, Sanitary Clamp to Male Thread, Straight, Avantor®
Fittings and Connectors
Maintain sanitary conditions while quickly and easily connecting to a male NPT thread.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
Tri-Clamp male thread adapters allow quick and easily installation of your sanitary set-up to an NPT male pipe thread. Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving. Clamps and gaskets required for a complete system are sold separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty