Would you like to log in again?
Masterflex® Concentric Reducer Fittings, Sanitary Clamp, Straight, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
TAD200-150 KAD150-075 KAD250-150 KAD200-100L KAD250-100L TAD300-250 TAD150-100L KAD150-075 KAD300-200 TAD250-200 KAD100L-075 KAD150-100L KAD200-150 TAD100L-075 KAD300-250 TAD250-150 TAD150-075 KAD250-150 TAD200-100L KAD250-200 KAD300-200 KAD200-150
MFLX40619-76EA 248.85 USD InStock
MFLX40619-76 MFLX40619-84 MFLX40619-85 MFLX40619-82 MFLX40619-83 MFLX40619-80 MFLX40619-81 MFLX31808-06 MFLX31808-18 MFLX31805-15 MFLX31808-15 MFLX31805-14 MFLX40619-79 MFLX31808-14 MFLX40619-77 MFLX31805-11 MFLX31805-10 MFLX40619-78 MFLX31805-17 MFLX31805-06 MFLX31805-05 MFLX31805-19
Masterflex® Concentric Reducer Fittings, Sanitary Clamp, Straight, Avantor®
Fittings and Connectors
Maintain sanitary conditions in your processes while decreasing diameter.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
Tri-Clamp® concentric reducers decrease in diameter towards the center thus forming a conical cylinder. Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving. Clamps and gaskets required for a complete system are sold separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty