Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Straight, HDPE, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
CO-2.5HDPE(100 PACK) C0-2-200HDPE-MF(10PK) C0-6HDPE(100 PACK) C0-2.5HDPE(10 PACK) C0-6HDPE(10 PACK) C0-1.5-200HDPE(100 PACK) C0-5HDPE(100 PACK) C0-4HDPE-MF(10PK) C0-10HDPE(10 PACK) C0-3HDPE-MF(10PK) C0-8HDPE-MF(10PK) C0-4HDPE(10 PACK) CO-3HDPE(100 PACK) C0-10HDPE-MF(10PK) C0-6HDPE-MF(10PK) C0-1-200HDPE-MF(10PK) C0-16HDPE-MF(10PK) C0-4HDPE(100 PACK) C0-1-200HDPE(10 PACK) C0-8HDPE(100 PACK) C0-2-200HDPE (10 PACK) C0-2-200HDPE(100 PACK) C0-1.5-200HDPE(10 PACK) C0-1-200HDPE(100 PACK) C0-5HDPE(10 PACK) C0-2-200HDPE(100 PACK) C0-12HDPE-MF(10PK) C0-3HDPE(10 PACK) C0-12HDPE(10 PACK) C0-8HDPE(10 PACK) C0-16HDPE(10 PACK)
MFLX40626-54EA 105 USD InStock
MFLX40626-54 MFLX30626-50 MFLX40626-53 MFLX30625-85 MFLX30612-09 MFLX30622-50 MFLX30625-87 MFLX30626-55 MFLX30625-89 MFLX40622-53 MFLX30622-55 MFLX30621-89 MFLX30621-87 MFLX30612-13 MFLX30621-85 MFLX30612-33 MFLX30621-83 MFLX30612-17 MFLX30621-81 MFLX40626-47 MFLX40626-44 MFLX40622-44 MFLX40622-47 MFLX30621-79 MFLX30612-21 MFLX30612-25 MFLX40612-01 MFLX30621-92 MFLX30612-29 MFLX30625-92 MFLX40612-05
Masterflex® Union Fittings, Hose Barb, Straight, HDPE, Avantor®
Fittings and Connectors
Quick and easy assembly without tools.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Ultra smooth inner surface results in fewer extractables
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty