Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Y-Connector, Polyproylene, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
222605 227065 227005 Y0-3 PP (100 PACK) Y0-2PP-QC-MF Y0-8 PP (100 PACK) Y0-8PP-MF(10PK) Y0-6PP-MF(10PK) Y0-6-AGPP Y0-8 PP (10 PACK) Y680-6005 (25PK) Y0-6 PP (100 PACK) Y0-3-AGPP Y0-1.5 PP (100 PACK) Y0-12PP (5 PACK) Y0-16PP (5 PACK) Y0-6 PP (10 PACK) Y0-4 PP (100 PACK) Y0-12PP-MF(10PK) 222555 Y0-2 PP (10 PACK) Y0-2 PP (100 PACK) Y0-1 PP (100 PACK) Y0-2-PP(10PK) Y0-4PP-MF(10PK) 222545 Y0-2-AGPP 222575 227025 Y0-4 PP (10 PACK) Y260-6005 (25PK) 227085 Y0-1PP-MF(10PK) Y670-6005 (25PK) Y0-10PP-MF(10PK) Y0-4-AGPP Y0-2PP-MF(10PK) Y0-1.5 PP (10 PACK) Y0-10 PP (10 PACK) Y230-6005 (25PK) Y6110-6005(25PK) Y0-6-PP(10PK) 222585 Y0-1 PP (10 PACK) 222595 227045 Y0-8-AGPP Y0-3PP-MF(10PK) Y0-3 PP (10 PACK) 222565
MFLX06295-70EA 56.3 USD InStock
MFLX06295-70 MFLX06368-18 MFLX06368-19 MFLX06295-30 MFLX06296-40 MFLX06296-00 MFLX30614-71 MFLX40728-43 MFLX41526-28 MFLX06295-60 MFLX40726-50 MFLX06295-20 MFLX30614-43 MFLX30614-47 MFLX06296-20 MFLX06295-50 MFLX06295-10 MFLX40617-80 MFLX40726-48 MFLX30614-51 MFLX40726-47 MFLX40726-46 MFLX40726-45 MFLX30614-55 MFLX40617-87 MFLX40726-44 MFLX40726-43 MFLX40726-42 MFLX40726-41 MFLX30614-59 MFLX40726-49 MFLX06295-40 MFLX40727-41 MFLX06296-50 MFLX06368-25 MFLX06296-10 MFLX06368-24 MFLX06368-22 MFLX30614-63 MFLX40617-79 MFLX40617-76 MFLX30614-67 MFLX40617-77 MFLX40727-42 MFLX41526-39 MFLX40727-43 MFLX40727-44 MFLX40727-45 MFLX40727-46 MFLX40727-47
Masterflex® Union Fittings, Hose Barb, Y-Connector, Polyproylene, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty