Would you like to log in again?
Masterflex® Reducer Fittings, Hose Barb, Straight (Polypropylene), Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
C4-2PP (100 PK) C12-6PP(50/PK) C10-8PP-MF C10-6PP(100/PK) C12-8PP(5 PACK) C10-8PP(10 PACK) C16-12PP(5 PACK) C4-3PP(10 PACK) C8-4PP(10 PACK) C10-8PP(100/PK) C3-2PP C1.5-1-200PP(250 PACK) C16-10PP(5 PACK) C2-1.5-200PP(25 PACK) C16-8PP(5 PACK) C8-4PP(100/PK) C2-1-200PP-MF C10-4PP(100/PK) C4-2.5PP (10PACK) C8-5PP(10 PACK) C2-1-200PP(25 PACK) C2-1-200PP(250 PACK) C12-6PP(5 PACK) C12-8PP(50/PK) C10-5PP(10 PACK) C16-8PP(50/PK) C16-10PP(50/PK) C6-5PP(10 PACK) C16-8PP-MF C2-1.5-200PP(250 PACK) C3-2PP-MF C8-6PP (10 PACK) C8-6PP(100/PK) C8-6PP-MF C1.5-1-200PP(25 PACK) C12-10PP(5 PACK) C2.5-2PP(10 PACK) C12-8PP-MF C8-5PP(100/PK) C4-3PP(100 PACK) C2-1.5-200PP-MF C6-4PP(10 PACK) C4-2PP C5-4PP(10 PACK) C6-5PP(100/PK) C4.2-5PP(100 PACK) C10-8PP (10 PACK) C6-4PP(100 PACK) C10-6PP(10 PACK) C10-4PP(10 PACK) C8-6PP(10 PACK)
MFLX41518-36EA 29.7 USD InStock
MFLX41518-36 MFLX41518-34 MFLX30616-51 MFLX50626-23 MFLX40610-18 MFLX50625-95 MFLX40610-19 MFLX50622-21 MFLX50626-21 MFLX50621-95 MFLX40640-08 MFLX40640-09 MFLX50622-23 MFLX40640-11 MFLX40640-12 MFLX40640-13 MFLX30616-63 MFLX40616-47 MFLX40610-20 MFLX40920-23 MFLX30616-67 MFLX40640-10 MFLX40616-43 MFLX40610-21 MFLX40610-22 MFLX40610-09 MFLX40626-32 MFLX40610-07 MFLX40610-08 MFLX40622-32 MFLX40640-15 MFLX40640-16 MFLX30616-71 MFLX40640-17 MFLX40640-18 MFLX30616-75 MFLX40640-20 MFLX40640-21 MFLX40610-12 MFLX40610-13 MFLX40610-10 MFLX40610-11 MFLX40610-16 MFLX40610-17 MFLX40610-14 MFLX40610-15 MFLX40626-26 MFLX40622-26 MFLX40622-19 MFLX30705-11 MFLX30705-12
Masterflex® Reducer Fittings, Hose Barb, Straight (Polypropylene), Avantor®
Fittings and Connectors
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range 9 to 180 °F (-13 to 66 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty