Would you like to log in again?
Union Fittings, Hose Barb Tee, Plastic
Supplier: VWR International
Masterflex®
T0-2-200NK7-MF 10PK T0-1.5-200BN T0-1-200HDPE T0-1-200BN T0-2PP-MF T0-2-200NK (100 PACK)) T0-4NK-MF T0-2-200AGHDPE (10 PACK)
MFLX40615-56EA 30.8 USD InStock
MFLX40615-56 MFLX40623-95 MFLX41519-56 MFLX40921-48 MFLX41519-54 MFLX41519-53 MFLX30613-47 MFLX30613-16
Union Fittings, Hose Barb Tee, Plastic
Fittings and Connectors
Designed for tight tolerances.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barb fittings make tubing connections quick and easy, requiring no tools for installation. Select the fitting based on the Masterflex tubing size. Fittings are available in a variety of materials to suit different applications and budgets.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty