Would you like to log in again?
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Nylon, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
FTLL230-2 (25PK) FTLL240-1 (25PK) FTLL004-1 (25PK) FTLL250-1 (25PK) FTLL013-1 (25PK) FTLL220-1 (25PK) FTLL055-1 (25PK) FTLL210-1 (25PK)
MFLX45502-02EA 17.9 USD InStock
MFLX45502-02 MFLX45502-01 MFLX45502-00 MFLX45502-20 MFLX45502-08 MFLX45502-06 MFLX45502-05 MFLX45502-04
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Nylon, Avantor®
Fittings and Connectors
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty