Would you like to log in again?
Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Nylon
Supplier: Value Plastics
FTLLB230-1 (1,000PK)
MFLX45502-34BLKEA 294 USD InStock
MFLX45502-34BLK
Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Nylon
Fittings and Connectors
Ensure your process integrity with full material traceability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty