Would you like to log in again?
Value Plastics® Adapter Fittings, Luer to Threaded
Supplier: Value Plastics
KFTLL-1 (1,000PK) XMTLL-1 (1,000PK) SFTLL-1 (1,000PK) SMTLL-1 (1,000PK) 14MTLL-1 (1,000PK) 18MTLL-1 (1,000PK) 18LFTLL-1 (1,000PK) 18FTLL-1 (1,000PK) BSFTLL-1 (1,000PK)
MFLX45505-86BLKEA 587 USD InStock
MFLX45505-86BLK MFLX45505-88BLK MFLX45505-76BLK MFLX45502-60BLK MFLX45505-84BLK MFLX45505-82BLK MFLX45505-80BLK MFLX45505-78BLK MFLX45502-66BLK
Value Plastics® Adapter Fittings, Luer to Threaded
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Easily complete your luer fittings systems with these premium connectors. Included are some special configurations available only in polycarbonate. The snap luer lock ring is for use with male luer by wide-bore hose barb fittings. Nylon material has good chemical resistance and can be sterilized via autoclave, ethylene oxide, or gamma irradiation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty