Would you like to log in again?
Masterflex® Adapter Fittings, Female Luer to Threaded, Straight, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
BSFTLL-6005 (25PK) KFTLL-9 (25PK) SFTLL-9 (25PK) KFTLL-6005 (25PK) SFTLL-6005 (25PK) 18FTLL-6005 (25PK)
MFLX45508-60EA 45.1 USD InStock
MFLX45508-60 MFLX45501-66 MFLX45508-66 MFLX45508-76 MFLX45501-60 MFLX30800-00
Masterflex® Adapter Fittings, Female Luer to Threaded, Straight, Avantor®
Fittings and Connectors
Easily transition between luer fittings and UNF or NPT threads.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty