Would you like to log in again?
Masterflex® Adapter Fittings, Hose Barb to Male UNF Threaded, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
A1032-1-200NN(25 PACK) B1 SS EA A1032-1.5-200PP(25 PACK) A1032-1.5-200HDPE(10 PACK) A1032T-1-200NK7-MF 10PK L1032T-2-200NK7-MF 10PK A1032-1.5-200NN(250 PACK) A1428-2-200NK7-MF 10PK A1032T-1.5-200BN(10PK) A1032-2-200HDPE(10 PACK) CGA48-PR0 A1032-1-200PP(250 PACK) A1032T-1.5-200HDPE(10P A1032-1-200HDPE(10 PACK) A1032-2-200NN(100 PACK) A1032T-2-200NK7-MF 10PK A1032T-1.5-200NK7-MF 10PK B3 SS EA CGA64-PR0 A1032-1.5-200HDPE(100 PACK) A1428-1.5NK7-MF 10PK A1032-1.5-200NN(25 PACK) A1032-1-200NN(250 PACK) CGA50-PR0 A1032-1.5-200PP(250 PACK) A1032-1-200HDPE(100 PACK) A1032-2-200NN(10 PACK) A1032T-2-200PP(10PK) CGA50-NR0 A1032-2-200HDPE(100 PACK) A1032T-1-200PP(10PK) A1032-2-200PP(25 PACK) A1032T-2-200HDPE(10PK) A1032-1-200PP(25 PACK)
MFLX40629-00EA 23.5 USD InStock
MFLX40629-00 MFLX40625-02 MFLX40629-01 MFLX50625-01 MFLX40625-00 MFLX40625-05 MFLX40614-91 MFLX40625-06 MFLX40621-00 MFLX40625-03 MFLX50621-01 MFLX50625-04 MFLX40621-02 MFLX41517-09 MFLX40621-03 MFLX50621-07 MFLX41517-04 MFLX40621-05 MFLX51517-03 MFLX50621-04 MFLX41517-05 MFLX40621-06 MFLX31209-40 MFLX31209-41 MFLX31500-55 MFLX51517-11 MFLX40614-00 MFLX40614-01 MFLX40614-02 MFLX31501-53 MFLX40614-03 MFLX40614-04 MFLX31501-54 MFLX31501-55
Masterflex® Adapter Fittings, Hose Barb to Male UNF Threaded, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty