Would you like to log in again?
Masterflex® Press-In Plug Fittings, Hose Barb, Straight, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
P0-1-200HDPE(10PK) P0-2.5-200NK7-MF 10PK P0-4NK (10 PACK) P0-1.5 200NK7-MF 10PK P0-1-200BN(10PK) P0-3-200NK7-MF 10PK PIP10-1 (10PK) P0-4NK7-MF 10PK P0-2-200PP(10PK) P0-1-200NK7-MF 10PK P0-2-200BN(10PK) P0-8NK7-MF 10PK P0-2-200NK7-MF 10PK P0-1-200PP(10PK) P0-6NK7-MF 10PK P0-2-200HDPE(10PK) P0-1.5BN
MFLX41519-36EA 18.2 USD InStock
MFLX41519-36 MFLX31220-11 MFLX41519-35 MFLX41519-44 MFLX31220-26 MFLX41519-43 MFLX51519-46 MFLX51519-38 MFLX31519-39 MFLX40615-44 MFLX40615-45 MFLX40615-40 MFLX40615-41 MFLX40615-42 MFLX40615-43 MFLX40615-38 MFLX40615-39
Masterflex® Press-In Plug Fittings, Hose Barb, Straight, Avantor®
Fittings and Connectors
Protect against contaminants in tubing lines.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Use these barbed press-in-plugs to keep open tubing lines or tubing assemblies free of foreign contaminants and ensure process purity. Plugs are manufactured from 100% virgin materials in a ISO Class 8 (formerly Class 100,000) environment and include full material traceability. Available in PVDF or nylon. All plugs are sterilizable by gamma irradiation; nylon plugs may also be autoclaved.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty