Would you like to log in again?
Masterflex® I/P® Easy-Load® Pump Head with BLDC Gearmotor and Controller, Panel Mount
Supplier: Avantor
Masterflex® I/P® Easy-Load®
900-2031
MFLX61047-78CS 2456 USD InStock
MFLX61047-78
Masterflex® I/P® Easy-Load® Pump Head with BLDC Gearmotor and Controller, Panel Mount
Pumps Custom Engineered Pumps
Versatile panel-mount peristaltic pump allow for simple integration into original equipment manufactured equipment where clean, low-shear, high flow, and high accuracy fluid dispensing is needed.
  • Compact design for efficient use of space
  • Three roller design provides positive displacement action without backflow
  • Automatic tubing retention eliminates user adjustments and makes setup easy - offers improved tubing life and improved performance
  • Accepts multiple tubing sizes for maximum flexibility-suitable for use with I/P® Precision pump tubing sizes (I/P 26, I/P 73, and I/P 82)
  • Brushless DC motor
Masterflex panel mount pump systems feature the flexibility of peristaltic pump technology and are designed for simple integration into system control panels for fluid transfer applications.
The Masterflex® I/P® Easy-Load® pump head combines an over-center cam with automatic tubing retention in a rugged, chemical-resistant housing. This design allows fast, easy tubing changes without removing the pump head from the drive and accommodates multiple tubing sizes. This is coupled to with a 24 V BLDC drive gearmotor with 0 - 5 VDC speed control, and direction control for precision control of process requirements.
The brushless DC motors design provides a low maintenance solution, perfect for applications where a long lifetime is needed.
Certifications: ROHS, REACH, ISO9001.
Ordering information: Order the I/P® Precision tubing size and formulation to suit your application (sold separately). More than 100 options exist - the most popular sizes and formulations can be found on the Accessory tab. Customizable options available, contact our technical support for more details.
Delivery information: Includes pump drive and pump head.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty