Would you like to log in again?
Masterflex® L/S® Easy-Load® Pump Head with BLDC Gearbox and Controller, Panel Mount
Supplier: Avantor
Masterflex® L/S® Easy-Load®
900-2445
MFLX61138-42CS 1708 USD InStock
MFLX61138-42
Masterflex® L/S® Easy-Load® Pump Head with BLDC Gearbox and Controller, Panel Mount
Pumps Custom Engineered Pumps
Versatile panel-mount peristaltic pumps allow for simple integration into original equipment manufactured equipment where clean, low-shear, and high accuracy fluid dispensing is needed.
  • Compact design for efficient use of space
  • Automatic tubing retention allow for multiple tubing sizes
  • Rotor is constructed of stainless steel (SS) for enhanced durability
  • Six-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Suitable for use with L/S® Precision pump tubing sizes (L/S 13, L/S 14, L/S 16)
  • Brushless DC motor
Masterflex panel mount pump systems feature the flexibility of peristaltic pump technology and are designed for simple integration into system control panels for fluid transfer applications.
The L/S® Easy-Load Pump Head included provides a simple, easy-to-use peristaltic pump system. The pump head accepts several different tubing sizes for a wide range of flow rates. The unique over-center cam design and automatic tubing retention allow quick tubing changes and greatly reduced maintenance time.
The brushless DC motors design provides a low maintenance solution, perfect for applications.
Certifications: ROHS, REACH, ISO9001.
Ordering information: Order the L/S® Precision tubing size and formulation to suit your application (sold separately). More than 100 options exist - the most popular sizes and formulations can be found on the Accessory tab. Customizable options available, contact our technical support for more details.
Delivery information: Includes pump drive and pump head.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty