Would you like to log in again?
Masterflex® Direct-Reading Variable-Area Flowmeters for Oxygen, Acrylic, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
ACRYS1V07 ACRZS1V07 ACRYB0V14 ACRZB1V14 ACRZB0V14 ACRYS0V07 ACRZS0V21 ACRYB1V07 ACRYB0V21 ACRZB0V07 ACRZS1V21 ACRYB1V21 ACRYS0V21 ACRYS0V28 ACRZS1V14 ACRYS0V14 ACRYS1V14 ACRZS0V14 ACRZB0V21 ACRYB1V28 ACRZB1V07 ACRYB0V07 ACRYB1V14 ACRZB1V21 ACRYS1V28 ACRYB0V28 ACRYS1V21
MFLX68560-36EA 74.3 USD InStock
MFLX68560-36 MFLX68560-37 MFLX68560-38 MFLX68560-39 MFLX68560-50 MFLX68560-51 MFLX68560-52 MFLX68560-30 MFLX68560-53 MFLX68560-31 MFLX68560-32 MFLX68560-54 MFLX68560-33 MFLX68560-55 MFLX68560-34 MFLX68560-47 MFLX68560-48 MFLX68560-49 MFLX68560-28 MFLX68560-29 MFLX68560-40 MFLX68560-41 MFLX68560-42 MFLX68560-43 MFLX68560-44 MFLX68560-45 MFLX68560-46
Masterflex® Direct-Reading Variable-Area Flowmeters for Oxygen, Acrylic, Avantor®
Flow Meters
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for oxygen. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty