Would you like to log in again?
Masterflex® Direct-Reading Variable-Area Flowmeters for Helium, Acrylic, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
ACRZB0V05 ACRYS1V24 ACRZB0V19 ACRZB1V12 ACRZS1V19 ACRYB0V19 ACRYS1V19 ACRYB0V26 ACRZS0V05 ACRYB1V26 ACRYB1V19 ACRYB1V12 ACRZS1V12 ACRZS0V19 ACRZB1V19 ACRYS0V12 ACRYS1V05 ACRZB0V12 ACRYB1V05 ACRZB1V05 ACRYS0V26 ACRYS0V05 ACRZS0V12 ACRYS1V12 ACRZS1V05
MFLX68561-48EA 65.1 USD InStock
MFLX68561-48 MFLX68561-49 MFLX68561-62 MFLX68561-63 MFLX68561-42 MFLX68561-64 MFLX68561-65 MFLX68561-43 MFLX68561-44 MFLX68561-66 MFLX68561-67 MFLX68561-68 MFLX68561-69 MFLX68561-47 MFLX68561-50 MFLX68561-59 MFLX68561-51 MFLX68561-52 MFLX68561-53 MFLX68561-55 MFLX68561-56 MFLX68561-57 MFLX68561-58 MFLX68561-60 MFLX68561-61
Masterflex® Direct-Reading Variable-Area Flowmeters for Helium, Acrylic, Avantor®
Flow Meters
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for helium. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty