Would you like to log in again?
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Supplier: BECKSON INDUSTRIAL PRODUCTS
CP3648S PC0960 CP1248S
MFLX70607-00EA 55 USD InStock
MFLX70607-00 MFLX70607-12 MFLX70607-10
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Pumps Drum/transfer Pumps
Ideal for most acid/base or ionic salt applications.
  • Portable and self-priming
These plastic hand-operated drum pumps are ideal for water or chemical siphons. All pumps are constructed with PVC barrel and hose. Water siphon model 70607-00 includes a 60” PVC outlet hose that can be cut to fit your application. Chemical siphon models 70607-10 and 70607-12 are suitable for acid and alkali transfer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty