Would you like to log in again?
Masterflex® Mechanically Coupled High-Head Centrifugal Pumps, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
ACDU200/115T1C A3U-32-12530T1C ACDU200/115T3C A3U-32-12530T3C A3U-32-160A50T3C A3U-40-16075T3C A3U-32-200-1100T3C A3U-32-20075T3C A3U-40-125B50T3C
MFLX70725-20EA 3230 USD InStock
MFLX70725-20 MFLX70725-31 MFLX70725-13 MFLX70725-11 MFLX70725-22 MFLX70725-32 MFLX70725-26 MFLX70725-37 MFLX70725-36
Masterflex® Mechanically Coupled High-Head Centrifugal Pumps, Avantor®
Pumps Centrifugal Pumps
Rugged continuous-duty service with chemical and corrosion resistance.
  • Close-coupled design saves space; simplifies maintenance and installation
  • Stainless steel liquid end components deliver corrosion resistance
  • Versatile mounting can be installed horizontally or vertically
  • Back-pullout construction assembly allows overhaul of impeller and seal without disturbing suction and discharge connections
  • Top centerline discharge and foot support under casing ensures self-venting and reduces misalignment from pipe loads
  • High operating efficiency lowers operating costs
The mechanically coupled centrifugal pumps, constructed of 304 stainless steel, are ideal for food and pharmaceutical processing plants, as well as water and wastewater treatment centers. They feature high-quality mechanical shaft seals and O-rings. Also available are optional materials for high-temperature and chemical-duty operations. Contact an Application Specialist for more information.
Delivery information: Includes conduit box for hard-wiring.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty