Would you like to log in again?
Masterflex® Flange Kits for Mechanically Coupled Centrifugal Pumps, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
3U-32 3U-40
MFLX70725-70EA 509 USD InStock
MFLX70725-70 MFLX70725-72
Masterflex® Flange Kits for Mechanically Coupled Centrifugal Pumps, Avantor®
Pumps Centrifugal Pumps
Enhance the life of your pump with service flange kits.
  • Constructed of stainless steel (SS) parts
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.
Delivery information: Includes stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty