Would you like to log in again?
Masterflex® Close-Coupled Industrial Gear Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
OBN999CP OBN991RCP OBN992RCP OBN992CP OBN991CP
MFLX70738-00EA 452 USD InStock
MFLX70738-00 MFLX70738-14 MFLX70738-04 MFLX70738-12 MFLX70738-02
Masterflex® Close-Coupled Industrial Gear Pump Heads, Avantor®
Pumps Pump Heads Gear Pump Heads
Rugged corrosion-resistant bronze construction provides long service life.
  • Easy assembly to NEMA and IEC motor frames
These gear pump heads are suitable for all liquids that are compatible with bronze—most common liquids are water, oil, and mild chemicals (pH from 4 to 11).
Choose a gear pump head to mount to your own motor (adapters for NEMA or IEC motors are sold separately below) or select a complete system with the pump head conveniently mounted to a standard (NEMA 56C) TEFC motor.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty