Would you like to log in again?
Polypropylene (PP) Moderate-Flow Chemical-Resistance Centrifugal Pumps
Supplier: BPH PUMP & EQUIPMENT INC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MD-20-220V MD-6 MD-40(T) MD-100RLT-115 MD-55(T) 230V MD-15 MD-30(T)-220V MD-100(T)-220V MD-70(T)-220V MD-10-220V MD-40(T)-220V MD-70(T) MD-30R(T) MD-55(T) MD-6-220V MD-10 MD-15-220V MD-20
MFLX72010-70EA 801 USD InStock
MFLX72010-70 MFLX72010-80 MFLX72010-65 MFLX72010-10 MFLX72010-20 MFLX72010-75 MFLX72010-45 MFLX72010-00 MFLX72010-55 MFLX72010-50 MFLX72010-60 MFLX72010-85 MFLX72010-30 MFLX72010-40 MFLX72010-25 MFLX72010-35 MFLX72010-05 MFLX72010-15
Polypropylene (PP) Moderate-Flow Chemical-Resistance Centrifugal Pumps
Pumps Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
Moderate-Flow Polypropylene Pumps are an ideal option for process, recirculation, or transfer applications where continuous flow and excellent chemical resistance are needed.
Delivery information: Includes a 6-ft (1.8 m) cord with stripped ends.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty