Would you like to log in again?
Polypropylene (PP) High-Head Chemical-Resistance Centrifugal Pumps
Supplier: BPH PUMP & EQUIPMENT INC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MD-70RZ(T)-220V MD-20Z(TUBE)-220V MD-20Z(TUBE) MD-30RZ(TUBE) MD-70RZ(T) MD-30Z(TUBE)-220V
MFLX72012-15EA 500 USD InStock
MFLX72012-15 MFLX72012-05 MFLX72012-25 MFLX72012-00 MFLX72012-20 MFLX72012-10
Polypropylene (PP) High-Head Chemical-Resistance Centrifugal Pumps
Pumps Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
High-head polypropylene pumps have a closed impeller design to significantly increase the output pressure. Polypropylene construction provides excellent chemical resistance in applications, such as filtration, where a high-head is needed.
Delivery information: Includes a 6-ft (1.8 m) cord with stripped ends.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty