Would you like to log in again?
Masterflex® Self-Priming Micro Pumps, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
130SP2440-1TP 130SP1240-1TP 130SP2430-1TP 130SP1220-1TP 130SP1230-1TP 120SP2430-4EE 120SP2420-4EE 120SP1250-4EE 120SP2440-4EE 150SP12250-4EE 130SP2420-1TP 150SP12100-4EE 120SP1220-4EE 150SP24250-4EE 120SP1230-4EE 120SP2450-4EE
MFLX73120-32EA 1170 USD InStock
MFLX73120-32 MFLX73120-10 MFLX73120-22 MFLX73120-30 MFLX73120-20 MFLX73120-64 MFLX73120-50 MFLX73120-40 MFLX73120-62 MFLX73120-38 MFLX73120-16 MFLX73120-36 MFLX73120-14 MFLX73120-34 MFLX73120-12 MFLX73120-24
Masterflex® Self-Priming Micro Pumps, Avantor®
Pumps Diaphragm Pumps
Accurate volumetric dispensing of 4 microliters to 250 microliters.
  • Positive shut with pressures less than 3 psi
  • High cycle life - up to 20 million dispense cycles
  • Low power consumption
These pumps are factory set for discrete outputs ranging from 4ìl to 250ìl. The pumps can be cycled at up to 2.0 Hz for the 73120-1* and -2* series of pumps. All the while the pump can maintain a high level of accuracy with a deviation from set-point being less than 1%.
The pumps are self-priming. Intially, the suction created by the larger pumps is sufficient to pull liquids from an unpressurized container located up to four feet below the. Once primed, the pump is able to maintain around 5 psi pressure.
Long life (3,000 hours) is built into each pump with a guarantee for up to 20 million actuations.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty