Would you like to log in again?
Micropump® B-Mount Gear Pump Heads
Supplier: Micropump
MI0016 / Z-140 / 82001 MI0006 / Z-186 / 82092 MI0267 / Z-040 / 81977 MI0013 / Z-120 / 82004 MI0007 / Z-181 / 82114 MI0131 / Z-1830 / 82004 MI0310 / Z-040HC / L24704 MI0278 / Z-186 PPS / L18412 MI0309 / Z-186HC / L2384/L2383 82006 MI0008 / Z-183 / 82115 MI0280 / Z-1830P / L18489 MI0313 / Z-140P / L19735 MI0312 / Z-186P / L20820 MI0264 / Z-050 / 81979 MI0019 / Z-130 / 81529 MI0018 / Z-142 / 82003 MI0020 / Z-150 / 81531
MFLX74004-71EA 2430 USD InStock
MFLX74004-71 MFLX74004-73 MFLX74004-13 MFLX74004-23 MFLX74004-01 MFLX74004-03 MFLX74004-53 MFLX74004-31 MFLX74004-75 MFLX74004-33 MFLX74004-55 MFLX74004-11 MFLX74004-09 MFLX74004-17 MFLX74004-27 MFLX74004-05 MFLX74004-07 MFLX74004-29
Micropump® B-Mount Gear Pump Heads
Pumps Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
  • Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Call our Application Specialists for assistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty