Would you like to log in again?
Masterflex® Digital Benchtop Gear Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
75211-70 & 74012-61 75211-70 & 74012-61 74014-32 75211-70 & 74012-11 75211-70 & 74012-51 75211-70 & 74012-51 75211-70 & 07002-25 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-51 74014-62 75211-70 & 74012-31 74014-12 75211-70 & 07002-25 75211-70 & 74012-11 75211-70 & 07002-14 75211-70 & 74012-21 74014-22 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 07003-04 75211-70 & 74012-31 75211-70 & 07002-25 74014-72 75211-70 & 74012-31 75211-70 & 74012-11 75211-70 & 74012-11 75211-70 & 74012-61 75211-70 & 07002-14 74014-42 75211-70 & 74012-31 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 74012-21 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 74012-51 74014-02 75211-70 & 07003-04 75211-70 & 74012-61 75211-70 & 74012-31 75211-70 & 74012-61 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 74012-11 75211-70 & 74012-51 75211-70 & 07003-04 75211-70 & 07002-14 75211-70 & 74012-61 74014-52 75211-70 & 07002-14 75211-70 & 07002-25 75211-70 & 74012-21 75211-70 & 74012-21 75211-70 & 74012-11 75211-70 & 07003-04 75211-70 & 74012-21 75211-70 & 74012-51 75211-70 & 07003-04 75211-70 & 07002-25 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 07002-14 75211-70 & 07002-25 75211-70 & 74012-61
MFLX74014-42-INCS 0 USD InStock
MFLX74014-42-IN MFLX74014-32-UK MFLX74014-52-EU MFLX74014-22-DK MFLX74014-42-IT MFLX74014-42-IL MFLX74014-52-AU MFLX74014-62-EU MFLX74014-42-UK MFLX74014-62-IT MFLX74014-02-DK MFLX74014-32-IT MFLX74014-72-CH MFLX74014-42 MFLX74014-12-DK MFLX74014-62-AU MFLX74014-32-IN MFLX74014-02 MFLX74014-32-IL MFLX74014-32-EU MFLX74014-22-CH MFLX74014-32-AU MFLX74014-62-IL MFLX74014-52-UK MFLX74014-62-IN MFLX74014-52-IL MFLX74014-72 MFLX74014-02-CH MFLX74014-32 MFLX74014-52-IT MFLX74014-42-EU MFLX74014-12-CH MFLX74014-52-IN MFLX74014-72-DK MFLX74014-62-UK MFLX74014-42-AU MFLX74014-42-CH MFLX74014-02-AU MFLX74014-62-DK MFLX74014-12-AU MFLX74014-12-EU MFLX74014-72-UK MFLX74014-72-IN MFLX74014-22-AU MFLX74014-72-IL MFLX74014-62 MFLX74014-52-DK MFLX74014-22-EU MFLX74014-32-CH MFLX74014-22 MFLX74014-22-IT MFLX74014-42-DK MFLX74014-72-IT MFLX74014-72-EU MFLX74014-02-UK MFLX74014-12-UK MFLX74014-22-IL MFLX74014-72-AU MFLX74014-22-IN MFLX74014-62-CH MFLX74014-02-IL MFLX74014-32-DK MFLX74014-02-IN MFLX74014-12-IT MFLX74014-52 MFLX74014-02-IT MFLX74014-12-IN MFLX74014-22-UK MFLX74014-12-IL MFLX74014-52-CH MFLX74014-12 MFLX74014-02-EU
Masterflex® Digital Benchtop Gear Pump Systems, Avantor®
Pumps Gear Pumps
Order the complete pump drive and head system.
  • Graphical LCD shows four operating modes
  • IP33 rating ensures splash and dust resistance
  • Systems deliver pulse-free flow in combination with Micropump® A-mount head
  • Remote control via DB25 female connector on drive
Calibrate, copy, and dispense features produce precise, pulseless batching and dispensing. View pump performance continually. Retrieve displacement and flow values by pump head size.
These precision systems are easily operated from the intuitive, multilanguage LCD front panel or optional foot switch 07523-92 for hands-free on/off control. The IP33-rated enclosure protects internal components from damage.
Service kits 07144-42 and 74012-52 include: gears, suction shoe, shaft seals, and springs
Service kit 74012-22 includes: gears, bushings, and seals
Delivery information: Includes 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country specific plug/cord set; please specify ultimate destination when ordering.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty