Would you like to log in again?
Pulsafeeder Economical Peristaltic Pumps
Supplier: EQUIP SOLUTIONS
XP015LAHX XP007LAHX XP030LALX XP004LFHX 7413024 XP100LVLX XP009LFLX XP050LFLX XP007LFHX
MFLX74130-12EA 631 USD InStock
MFLX74130-12 MFLX74130-02 MFLX74130-24 MFLX74130-13 MFLX74130-11 MFLX74130-04 MFLX74130-26 MFLX74130-07 MFLX74130-18
Pulsafeeder Economical Peristaltic Pumps
Pumps Peristaltic Pumps
Easy maintenance and no contamination—tubing is the only wetted part.
  • Easy tube change-out minimizes downtime between tubing set changes
  • Inherent degassing function and rugged gear train offer many years of reliable service
These simple-to-use, self-priming peristaltic pumps deliver worry-free dosing.
Delivery information: Includes pump head, two tubing assemblies, and a 6-ft (1.8-m) power cord.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty