Would you like to log in again?
LMI Manually Adjustable Control Metering Pumps
Supplier: LMI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
W2T464407 W2T464932 C131-318SI W2T458030 B121-398SI
MFLX74513-25EA 1569 USD InStock
MFLX74513-25 MFLX74513-20 MFLX74513-30 MFLX74513-40
LMI Manually Adjustable Control Metering Pumps
Pumps Diaphragm Pumps
Manually adjustable stroke length and stroke frequency provide precise control of flow rates.
  • Clear face cover protects pump controls on low flow models (1 GPH and lower)
  • Chemical-resistant housing provides protection in harsh environments
These pumps include a four-function valve — an anti-siphon, back pressure, priming, and pressure relief valve all in one.
What's included: a four-function valve, 16-ft polyethylene tubing, foot valve/strainer, and 8-ft power cord with molded plug.
Service kits include a replacement diaphragm, seal, four pump head screws, and check valves.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty