Would you like to log in again?
LMI Microprocessor-Controlled Solenoid Diaphragm Metering Pumps
Supplier: LMI
AD961-948SI AD811-918SI AD255-938SI AD241-928SI AD841-928SI AD951-938SI AD211-918SI AD851-938SI AD261-948SI AD245-928SI AD911-918SI AD861-948SI AD941-928SI AD251-938SI
MFLX74522-42EA 1953 USD InStock
MFLX74522-42 MFLX74522-22 MFLX74522-44 MFLX74522-24 MFLX74522-46 MFLX74522-02 MFLX74522-14 MFLX74522-48 MFLX74522-04 MFLX74522-26 MFLX74522-16 MFLX74522-28 MFLX74522-06 MFLX74522-08
LMI Microprocessor-Controlled Solenoid Diaphragm Metering Pumps
Pumps Diaphragm Pumps
Robust processor-based control offers improved electromagnetics.
  • Easy-to-use push button controls with LCD user interface
  • Manual stroke length knob
  • Low level float switch connection
  • Lockable clear cover
  • NEMA 4X / IP65 enclosure
Choose from manual control, remote control, and digital remote control pump options.
Manual control models feature manual stroke length and frequency adjustment. Low level indication is possible via the remote input. LCD provides quick user feedback of pump status and theoretical pump flow.
Remote control models accept a pulse or 4 to 20 mA input control signal. Use the keypad to manually adjust the stroke length and frequency (when in local mode). Pump has a dual low-level float switch input and a 24 V output. LCD provides quick user feedback of pump status and theoretical pump flow.
Digital remote control models offer all the same features of the remote control pumps as well as remote on/off and remote internal/external inputs; plus, 4 to 20 mA, pulse, and alarm outputs. An upgraded LCD offers a graphical display of pump status and theoretical pump flow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty