Would you like to log in again?
Suction Line Pump Filter Strainers
Supplier: SPECIALTY MFG CO
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3991570 3990680 3990550 3990610 3991650 3991560 3991550 3990980 3990560 3990570 3990540 3991640 3990600 3991630 3990670 3991350 3990750 3991500 3991320 3991130
MFLX74604-52EA 25.1 USD InStock
MFLX74604-52 MFLX74604-30 MFLX74604-42 MFLX74604-53 MFLX74604-20 MFLX74604-23 MFLX74604-12 MFLX74604-34 MFLX74604-24 MFLX74604-02 MFLX74604-32 MFLX74604-54 MFLX74604-43 MFLX74604-10 MFLX74604-44 MFLX74604-22 MFLX74604-33 MFLX74605-12 MFLX74604-14 MFLX74604-04
Suction Line Pump Filter Strainers
Filters Strainers
Protect pump from sediments filtering into suction line.
  • PVC (polyvinyl chloride) or Nylon base material of construction
  • 20, 40, 80 mesh sizes available
  • Easily removable for cleaning or replacing
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty