Would you like to log in again?
Masterflex® Dual-Drive Syringe Pump Accessories, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
78-0393 78-0394
MFLX74901-88EA 215 USD InStock
MFLX74901-88 MFLX74901-87
Masterflex® Dual-Drive Syringe Pump Accessories, Avantor®
Pumps Syringe Pumps
Daisy chain up to 100 pumps.
  • Connect multiple independent syringes to operate simultaneously
  • Easily connect pumps
  • Choose cable based on your current pump
Optimize performance of your Masterflex® Dual-Drive Syringe Pump with these accessories.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty