Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Touch-Screen, Continuous-Cycle, Four-Syringe Pumps, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
788272C 788270C
MFLX74905-39EA 6120 USD InStock
MFLX74905-39 MFLX74905-19
Masterflex® Touch-Screen, Continuous-Cycle, Four-Syringe Pumps, Avantor®
Pumps Syringe Pumps
Continuously cycle up to four syringes in a push-pull action.

  • Flow range from 5 pL/min to 215.8 ml/min
  • Intuitive touch screen allows you to quickly create and recall configurations
  • You can daisy chain up to 99 pumps together using the built in RS-485
  • Accuracy of ±0.35% and reproducibility of ±0.05%

Hold up to four syringes to cycle continuously back and forth in a push-pull action. As two syringes are infusing, two syringes are simultaneously withdrawing at the same rate. At the end of the set volume the direction is automatically reversed and the n

Delivery information: Includes 6 ft (1.8 m) U.S. power cord.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty