Would you like to log in again?
Multi-Position Double-Diaphragm Pumps
Supplier: XYLEM - FLOW CONTROL
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
D3835H5011A D3835E7011A D3935H5011A
MFLX75000-10EA 407 USD InStock
MFLX75000-10 MFLX75000-00 MFLX75000-05
Multi-Position Double-Diaphragm Pumps
Pumps Diaphragm Pumps
No metal wetted parts permit use with corrosive liquids.
  • All models feature factory-set pressure limits to protect pumps from damage—pumping stops when high limit is reached, resumes when pressure decreases to low setting.
Pump head rotates and mounts in 90° increments—position ports in one of four orientations to simplify installation. Model 75000-20 has a four-chamber design for higher flow rates and smoother flow. Pumps for 115 VAC operation include 6-ft power cord with three-prong plug. Order optional inlet filter under accessories to filter particulates.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty