Would you like to log in again?
Warren Rupp Surge Suppressors and Dampeners
Supplier: Warren Rupp
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DA071PPTNS00 DA071KPTNS00 DA051PPTNS00 TA1-1/2-NG1A DA101PPTAS00 DA051PPTNS00 TA1-NG1A
MFLX75008-12EA 1197 USD InStock
MFLX75008-12 MFLX75008-00 MFLX75008-02 MFLX75008-30 MFLX75008-40 MFLX75008-10 MFLX75008-20
Warren Rupp Surge Suppressors and Dampeners
Pumps Diaphragm Pumps
Minimize pulsation of positive displacement pumps.
  • Less vibration and noise
  • Simple installation
  • Variety of materials of construction to meet the system conditions
These Warren Rupp surge suppressors and dampeners are ideal for use to minimize pulsation while pumping with your positive displacement pump. Choose from a wide range of materials based on the chemical compatibility required.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty