Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Masterflex® Console Gear Pump Drive for MicroPump A-Mount Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
75211-12 75211-17 75211-17 75211-17 75211-17 75211-17 75211-17 75211-17 75211-17 75211-17
MFLX75211-17-DKEA 1570 USD InStock
MFLX75211-17-DK MFLX75211-17-IT MFLX75211-17 MFLX75211-17-EU MFLX75211-17-IL MFLX75211-12 MFLX75211-17-AU MFLX75211-17-CH MFLX75211-17-IN MFLX75211-17-UK
Masterflex® Console Gear Pump Drive for MicroPump A-Mount Pump Heads, Avantor®
Pumps Pump Drives Gear Pump Drives
Simple variable speed control handles many fluid handling applications.
  • 100:1 speed-control range controls flow over a wide range
  • Maximum continuous-duty flow rates possible with Micropump® A-mount pump heads
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Couple with an A-mount gear pump head for pulseless fluid flow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty