Would you like to log in again?
Masterflex® FDA Compliant Sanitary Drum Pumps, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
SP-8600-39 W/ENC-2-V SP-8600-39 W/ SP-280P-2-V SP-8700-39 W/ SP-A2 SP-8600-39 W/ SP-ENC-V SP-8700-39 W/ SP-ENC-2-V SP-8600-39 W/ SP-A2 SP-8700-39 W/ SP-280P-V SP-8600-39 W/ SP-280P-V SP-8700-39 W/ SP-ENC-V
MFLX75219-10EA 3650 USD InStock
MFLX75219-10 MFLX75219-20 MFLX75219-55 MFLX75219-00 MFLX75219-50 MFLX75219-60 MFLX75219-40 MFLX75219-05 MFLX75219-15
Masterflex® FDA Compliant Sanitary Drum Pumps, Avantor®
Pumps Drum/transfer Pumps
Capable of pumping fluids with viscosities up to 1000 cps.
  • 316 SS construction with PTFE elastomers
  • Easy to clean with Tri-Clamp® fittings
The sanitary drum pumps are engineered to transfer low-viscosity food products, pharmaceutical materials, and personal care solutions from drums, totes, and kettles. Wetted surfaces are constructed with FDA-compliant materials which provide an ideal solution for sanitary applications.
Delivery information: Includes drive and pump tube.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty